The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109661.1 | NP_001103131.1 | 3 - 4 | 1993 | 91 |
GenBank mRNA | BC131642.1 | - | 3 - 4 | 1901 | 91 |