Product Details
- Assay Reference Genome
Location
Chr.17:55954473 on build GRCm38
- Cytoband
- 17D
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 50498
- Gene Symbol
- Ebi3
- Gene Name
- Epstein-Barr virus induced gene 3
- Gene Aliases
- EBI-3, IL-27
- Location
- Chr.17:55952623-55957022 on build GRCm38
- Assay Gene Location
- Overlaps Exon 3 - Intron 3
- Entrez Gene ID
- 240120
- Gene Symbol
- Zfp119b
- Gene Name
- zinc finger protein 119b
- Gene Aliases
- Location
- Chr.17:55938373-55956178 on build GRCm38
- Assay Gene Location
Gene Symbol |
Transcript Accession |
Exon Location |
Assay Location |
Protein ID |
Zfp119b |
|
|
|
|
View Full Product Details