The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133768.4 | NP_598529.1 | 16 - 17 | 1334 | 73 |
GenBank mRNA | AK010922.1 | - | 16 - 17 | 1346 | 73 |
AK076453.1 | - | 16 - 17 | 1349 | 73 | |
AK138423.1 | - | 16 - 17 | 1331 | 73 | |
AK140160.1 | - | 16 - 17 | 1349 | 73 | |
AK148764.1 | - | 16 - 17 | 1318 | 73 | |
AK168739.1 | - | 16 - 17 | 1329 | 73 | |
BC016670.1 | - | 16 - 17 | 1334 | 73 |