Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nags
Entrez Gene ID
217214
Gene Name
N-acetylglutamate synthase
Gene Aliases
1700120E20Rik, argA
Chromosome Location
Chr.11: 102145513 - 102149528 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145829.2 NP_665828.1 3 - 4 972 54
GenBank mRNA AF462069.1 - 3 - 4 928 54
AJ489814.1 - 3 - 4 898 54
AK075765.1 - 2 - 3 618 54
BC057990.1 - 3 - 4 936 54

View Full Product Details