Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrn1
Entrez Gene ID
68404
Gene Name
neuritin 1
Gene Aliases
0710008J23Rik, Cpg15, Nrn
Chromosome Location
Chr.13: 36725622 - 36734477 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153529.2 NP_705757.1 1 - 2 245 88
GenBank mRNA AK003046.1 - 1 - 2 72 88
AK161859.1 - 1 - 2 213 88
BC035531.1 - 1 - 2 233 88

View Full Product Details