The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286411.1 | NP_001273340.1 | 2 - 3 | 399 | 100 |
NM_001286412.1 | NP_001273341.1 | 2 - 3 | 399 | 100 | |
NM_007920.4 | NP_031946.1 | 2 - 3 | 746 | 100 | |
GenBank mRNA | AK132456.1 | - | 2 - 3 | 400 | 100 |
AK156098.1 | - | 2 - 3 | 389 | 100 | |
BC027532.1 | - | 2 - 3 | 367 | 100 | |
BC057134.1 | - | 2 - 3 | 746 | 100 | |
U19617.1 | - | 2 - 3 | 393 | 100 |