The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146348.1 | NP_001139820.1 | 4 - 5 | 891 | 77 |
NM_001146350.1 | NP_001139822.1 | 4 - 5 | 894 | 77 | |
NM_007932.2 | NP_031958.2 | 4 - 5 | 894 | 77 | |
GenBank mRNA | AK151302.1 | - | 4 - 5 | 864 | 77 |
AK178235.1 | - | 2 - 3 | 194 | 77 | |
AK188120.1 | - | 2 - 3 | 194 | 77 | |
AY679531.1 | - | 4 - 5 | 894 | 77 | |
BC029080.1 | - | 4 - 5 | 808 | 77 | |
S69407.1 | - | 5 - 6 | 759 | 77 | |
X77952.1 | - | 4 - 5 | 894 | 77 |