Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn5
Entrez Gene ID
13863
Gene Name
lipocalin 5
Gene Aliases
E-RABP, Erabp, MEP10
Chromosome Location
Chr.2: 25657932 - 25662230 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042630.2 NP_001036095.1 3 - 4 361 68
NM_001276257.1 NP_001263186.1 3 - 4 361 68
NM_007947.3 NP_031973.2 3 - 4 361 68
GenBank mRNA AK021014.1 - 3 - 4 359 68
AK136629.1 - 3 - 4 364 68
AK162355.1 - 3 - 4 360 68
U68546.1 - 3 - 4 257 68

View Full Product Details