The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127355.1 | NP_001120827.1 | 9 - 10 | 1149 | 65 |
NM_001127356.1 | NP_001120828.1 | 8 - 9 | 1371 | 65 | |
NM_010122.2 | NP_034252.2 | 9 - 10 | 1004 | 65 | |
GenBank mRNA | AK158282.1 | - | 9 - 10 | 913 | 65 |
BC021884.1 | - | 9 - 10 | 891 | 65 | |
M98035.1 | - | 8 - 9 | 991 | 65 | |
M98036.1 | - | 9 - 10 | 957 | 65 |