The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011729.2 | NP_035859.2 | 1 - 2 | 404 | 65 |
GenBank mRNA | AK038822.1 | - | 1 - 2 | 233 | 65 |
AK137563.1 | - | 1 - 2 | 236 | 65 | |
AK164205.1 | - | 1 - 2 | 222 | 65 | |
BC053034.1 | - | 1 - 2 | 213 | 65 | |
BC092235.1 | - | 1 - 2 | 193 | 65 | |
BC141070.1 | - | 1 - 2 | 234 | 65 | |
D16306.1 | - | 1 - 2 | 99 | 65 | |
U40796.1 | - | 1 - 2 | 300 | 65 |