The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015779.2 | NP_056594.2 | 4 - 5 | 621 | 107 |
GenBank mRNA | AK143710.1 | - | 4 - 5 | 621 | 107 |
BC145800.1 | - | 4 - 5 | 686 | 107 |