Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exoc7
Entrez Gene ID
53413
Gene Name
exocyst complex component 7
Gene Aliases
Exo70, sec70
Chromosome Location
Chr.11: 116287998 - 116306749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162872.1 NP_001156344.1 4 - 5 481 57
NM_016857.2 NP_058553.2 4 - 5 481 57
GenBank mRNA AF014461.1 - 4 - 5 416 57
AK087601.1 - 4 - 5 469 57
AK140450.1 - 4 - 5 455 57
AK162868.1 - 4 - 5 467 57
AK167769.1 - 4 - 5 464 57
AK169928.1 - 4 - 5 468 57
AK173094.1 - 4 - 5 457 57
BC028927.1 - 4 - 5 457 57
BC053710.1 - 4 - 5 468 57

View Full Product Details