The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022983.4 | NP_075359.1 | 2 - 3 | 948 | 88 |
GenBank mRNA | AF293845.2 | - | 2 - 3 | 913 | 88 |
AK042084.1 | - | 2 - 3 | 925 | 88 | |
AK042255.1 | - | 2 - 3 | 937 | 88 | |
AK132289.1 | - | 2 - 3 | 891 | 88 | |
BC125460.1 | - | 1 - 2 | 766 | 88 | |
BC125462.1 | - | 1 - 2 | 766 | 88 |