Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
67426
Gene Name
aarF domain containing kinase 3
Gene Aliases
4632432J16Rik, AI462003, Cabc1, mKIAA0451
Chromosome Location
Chr.1: 180165238 - 180196020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163290.1 NP_001156762.1 5 - 6 774 69
NM_023341.3 NP_075830.2 5 - 6 872 69
GenBank mRNA AJ278735.1 - 5 - 6 754 69
AK004791.1 - 5 - 6 814 69
AK014605.2 - 5 - 6 814 69
AK178973.1 - 3 - 4 275 69
AK207690.1 - 3 - 4 276 69
BC030937.1 - 5 - 6 820 69
U31629.1 - 3 - 4 413 69

View Full Product Details