Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfcp2l1
Entrez Gene ID
81879
Gene Name
transcription factor CP2-like 1
Gene Aliases
1810030F05Rik, 4932442M07Rik, AA575098, Cp2l1, Crtr-1, D930018N21Rik, LBP-9, Tcfcp2l1
Chromosome Location
Chr.1: 118627923 - 118685168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023755.2 NP_076244.2 4 - 5 491 106
GenBank mRNA AF311309.1 - 4 - 5 486 106
AK143960.1 - 4 - 5 490 106
BC116663.1 - 4 - 5 486 106

View Full Product Details