The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024217.3 | NP_077179.1 | 2 - 3 | 459 | 67 |
GenBank mRNA | AK003882.1 | - | 2 - 3 | 314 | 67 |
AK079165.1 | - | 2 - 3 | 428 | 67 | |
AK172091.1 | - | 2 - 3 | 362 | 67 | |
AY241870.1 | - | 2 - 3 | 453 | 67 | |
BC003230.1 | - | 2 - 3 | 366 | 67 | |
BC145817.1 | - | 2 - 3 | 331 | 67 | |
BC145819.1 | - | 2 - 3 | 331 | 67 |