Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp16
Entrez Gene ID
74112
Gene Name
ubiquitin specific peptidase 16
Gene Aliases
1200004E02Rik, 2810483I07Rik, 6330514E22Rik, UBPM
Chromosome Location
Chr.16: 87454708 - 87483515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024258.2 NP_077220.2 14 - 15 2179 69
GenBank mRNA AK004594.2 - 14 - 15 2154 69
AK013437.1 - 8 - 9 1195 69
AK160038.1 - 14 - 15 2149 69
BC003278.1 - 14 - 15 2108 69
BC004577.1 - 2 - 3 691 69

View Full Product Details