The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026156.2 | NP_080432.1 | 6 - 7 | 845 | 97 |
GenBank mRNA | AK002890.1 | - | 6 - 7 | 846 | 97 |
AK031533.1 | - | - | 855 | 97 | |
AK087952.1 | - | 6 - 7 | 852 | 97 | |
AK132434.1 | - | 6 - 7 | 859 | 97 | |
AK145925.1 | - | 6 - 7 | 847 | 97 | |
BC021341.1 | - | 6 - 7 | 843 | 97 |