Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eepd1
Entrez Gene ID
67484
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
2310005P05Rik, AI481005
Chromosome Location
Chr.9: 25481526 - 25604110 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026189.3 NP_080465.3 2 - 3 1341 69
GenBank mRNA AK009180.2 - 2 - 3 1336 69
AK019086.1 - 2 - 3 1336 69
AK168564.1 - 2 - 3 1342 69
AK173239.1 - 2 - 3 1391 69
BC025571.1 - 2 - 3 1405 69
CK619108.1 - 1 - 2 69 69

View Full Product Details