The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142681.1 | NP_001136153.1 | 2 - 3 | 239 | 85 |
NM_026522.5 | NP_080798.2 | 2 - 3 | 239 | 85 | |
GenBank mRNA | AK014071.1 | - | 2 - 3 | 188 | 85 |
AK014141.1 | - | 2 - 3 | 214 | 85 | |
AK036717.1 | - | 2 - 3 | 183 | 85 | |
AK050655.1 | - | 2 - 3 | 187 | 85 | |
AK147377.1 | - | 2 - 3 | 202 | 85 | |
AK160156.1 | - | 2 - 3 | 199 | 85 | |
AK160200.1 | - | 2 - 3 | 199 | 85 | |
BC006876.1 | - | 2 - 3 | 188 | 85 | |
BC061063.1 | - | 2 - 3 | 199 | 85 | |
BY101128.1 | - | 2 - 3 | 240 | 85 |