The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027019.3 | NP_081295.2 | 5 - 6 | 564 | 72 |
GenBank mRNA | AB067773.1 | - | 5 - 6 | 653 | 72 |
AK005883.1 | - | 5 - 6 | 675 | 72 | |
BC104105.1 | - | 5 - 6 | 564 | 72 | |
BC104106.1 | - | 5 - 6 | 564 | 72 | |
BC106806.1 | - | 5 - 6 | 564 | 72 |