The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285905.1 | NP_001272834.1 | 3 - 4 | 399 | 76 |
NM_001285906.1 | NP_001272835.1 | 3 - 4 | 413 | 76 | |
NM_001285907.1 | NP_001272836.1 | 3 - 4 | 541 | 76 | |
NM_027353.4 | NP_081629.1 | 3 - 4 | 393 | 76 | |
GenBank mRNA | AK010582.1 | - | 3 - 4 | 278 | 76 |
AK031694.1 | - | 3 - 4 | 307 | 76 | |
AK034689.1 | - | 3 - 4 | 317 | 76 | |
AK154346.1 | - | 3 - 4 | 278 | 76 | |
AK164102.1 | - | 2 - 3 | 257 | 76 | |
AK167249.1 | - | 3 - 4 | 339 | 76 | |
AK173122.1 | - | 2 - 3 | 235 | 76 | |
BC006920.1 | - | 3 - 4 | 309 | 76 | |
BC079575.1 | - | 2 - 3 | 467 | 76 | |
BM944739.1 | - | 3 - 4 | 323 | 76 |