Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Speer4f
Entrez Gene ID
70935
Gene Name
spermatogenesis associated glutamate (E)-rich protein 4f
Gene Aliases
4922502J04Rik, SPEER-4F
Chromosome Location
Chr.5: 17476098 - 17480936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027609.2 NP_081885.1 2 - 3 292 91
GenBank mRNA AK015022.1 - 2 - 3 295 91
BC116901.1 - 2 - 3 318 91
BC116903.1 - 2 - 3 318 91

View Full Product Details