Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep89
Entrez Gene ID
72140
Gene Name
centrosomal protein 89
Gene Aliases
2610507L03Rik, Ccdc123
Chromosome Location
Chr.7: 35397000 - 35439828 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028120.2 NP_082396.1 18 - 19 2205 83
GenBank mRNA AK012067.1 - 18 - 19 2206 83
AK029107.1 - 18 - 19 2236 83
BC003981.1 - 19 - 20 2457 83

View Full Product Details