The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160211.1 | NP_001153683.1 | 5 - 6 | 1251 | 95 |
NM_028408.3 | NP_082684.1 | 5 - 6 | 1308 | 95 | |
GenBank mRNA | AK013789.1 | - | 5 - 6 | 1305 | 95 |
AK033911.1 | - | 5 - 6 | 1249 | 95 | |
BC115640.1 | - | 5 - 6 | 877 | 95 | |
BC115641.1 | - | 5 - 6 | 877 | 95 |