The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030719.3 | NP_109644.2 | 3 - 4 | 671 | 73 |
GenBank mRNA | AJ296173.1 | - | 3 - 4 | 711 | 73 |
AK039122.1 | - | 3 - 4 | 697 | 73 | |
AK052217.1 | - | 3 - 4 | 656 | 73 | |
BC094613.1 | - | 3 - 4 | 671 | 73 |