Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pramel1
Entrez Gene ID
83491
Gene Name
preferentially expressed antigen in melanoma-like 1
Gene Aliases
-
Chromosome Location
Chr.4: 143394359 - 143400160 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031377.2 NP_113554.1 2 - 3 421 70
GenBank mRNA AF285578.1 - 2 - 3 456 70
BC131995.1 - 2 - 3 422 70
BC132399.1 - 2 - 3 422 70

View Full Product Details