The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031392.2 | NP_113569.1 | 5 - 6 | 2993 | 91 |
GenBank mRNA | AF348591.1 | - | 5 - 6 | 2989 | 91 |
AK037181.1 | - | 5 - 6 | 2970 | 91 | |
AK046119.1 | - | 5 - 6 | 2961 | 91 | |
AK052235.1 | - | 5 - 6 | 2934 | 91 | |
AK146617.1 | - | 5 - 6 | 2950 | 91 | |
AK163752.1 | - | 5 - 6 | 2926 | 91 | |
AK165935.1 | - | 5 - 6 | 2957 | 91 | |
AK166265.1 | - | 5 - 6 | 2971 | 91 | |
AK169357.1 | - | 5 - 6 | 2994 | 91 | |
AK177316.1 | - | 2 - 3 | 173 | 91 | |
AK187031.1 | - | 2 - 3 | 175 | 91 | |
AK215607.1 | - | 2 - 3 | 174 | 91 | |
BC035951.1 | - | 4 - 5 | 972 | 91 | |
BC050894.1 | - | 5 - 6 | 2993 | 91 | |
BC054367.1 | - | 5 - 6 | 2921 | 91 |