Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Entpd7
- Entrez Gene ID
-
93685
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 7
- Gene Aliases
-
1810012B13Rik, 1810020C02Rik, 2810003F23Rik, LALP1, Lysal2
- Chromosome Location
-
Chr.19: 43689689 - 43733853 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF288221.1 |
- |
|
764 |
|
Chr. - - [-] on Build |
103 |
View Full Product Details