The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053130.3 | NP_444360.1 | 1 - 1 | 229 | 93 |
GenBank mRNA | AK028661.1 | - | 1 - 1 | 231 | 93 |
AK049825.1 | - | 1 - 1 | 219 | 93 | |
AY013787.1 | - | 1 - 1 | 41 | 93 |