Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim11
Entrez Gene ID
94091
Gene Name
tripartite motif-containing 11
Gene Aliases
-
Chromosome Location
Chr.11: 58978075 - 58991462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053168.2 NP_444398.1 3 - 4 879 80
GenBank mRNA AF220124.1 - 3 - 4 856 80
AK040618.1 - 3 - 4 880 80
AK051266.1 - 3 - 4 838 80
AK139477.1 - 3 - 4 864 80
AK157332.1 - 3 - 4 872 80
AK170656.1 - 3 - 4 845 80
AK170855.1 - 3 - 4 873 80

View Full Product Details