Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpsf1
Entrez Gene ID
94230
Gene Name
cleavage and polyadenylation specific factor 1
Gene Aliases
-
Chromosome Location
Chr.15: 76595808 - 76607613 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164173.1 NP_001157645.1 25 - 26 3084 59
NM_053193.2 NP_444423.1 25 - 26 3084 59
GenBank mRNA AF322193.1 - 25 - 26 3055 59
AK149914.1 - 18 - 19 2083 59
AK155806.1 - 25 - 26 3006 59
BC011458.1 - 21 - 22 2533 59
BC049941.1 - 25 - 26 2906 59
BC056388.1 - 25 - 26 3084 59
BC057945.1 - 10 - 11 1415 59

View Full Product Details