Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxp2
Entrez Gene ID
114142
Gene Name
forkhead box P2
Gene Aliases
2810043D05Rik, AI449000, CAG-16, D0Kist7
Chromosome Location
Chr.6: 14901226 - 15441977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286607.1 NP_001273536.1 10 - 11 1522 62
NM_053242.4 NP_444472.2 12 - 13 1922 62
NM_212435.1 NP_997600.1 10 - 11 1585 62
GenBank mRNA AF339106.1 - 9 - 10 1263 62
AK051668.1 - 12 - 13 1905 62
AK082361.1 - 12 - 13 1594 62
AY079003.1 - 12 - 13 1551 62
BC058960.1 - 10 - 11 1519 62
BC062926.1 - 10 - 11 1585 62

View Full Product Details