The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039077.2 | NP_001034166.1 | 4 - 5 | 379 | 79 |
NM_001039078.2 | NP_001034167.1 | 3 - 4 | 343 | 79 | |
NM_054057.4 | NP_473398.1 | 4 - 5 | 379 | 79 | |
GenBank mRNA | AK032308.1 | - | 4 - 5 | 329 | 79 |
AK134549.1 | - | 4 - 5 | 312 | 79 | |
AK147670.1 | - | 4 - 5 | 361 | 79 | |
AK160172.1 | - | 4 - 5 | 333 | 79 | |
BC047992.1 | - | 3 - 4 | 300 | 79 | |
BC061045.1 | - | 4 - 5 | 373 | 79 |