Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adcy4
Entrez Gene ID
104110
Gene Name
adenylate cyclase 4
Gene Aliases
mKIAA4004
Chromosome Location
Chr.14: 55769056 - 55784095 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080435.1 NP_536683.1 25 - 26 3195 55
GenBank mRNA AF442771.1 - 25 - 26 3195 55
AK141767.1 - 25 - 26 3266 55
AK161880.1 - 25 - 26 3214 55
AK220443.1 - 20 - 21 4075 55
BC015299.1 - 25 - 26 3195 55

View Full Product Details