The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080444.4 | NP_536692.1 | 2 - 3 | 683 | 87 |
GenBank mRNA | AF398971.1 | - | 2 - 3 | 712 | 87 |
AK028538.1 | - | 2 - 3 | 683 | 87 | |
AK052929.1 | - | 2 - 3 | 599 | 87 | |
BC141311.1 | - | 2 - 3 | 674 | 87 | |
BC141313.1 | - | 2 - 3 | 674 | 87 |