Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asb10
Entrez Gene ID
117590
Gene Name
ankyrin repeat and SOCS box-containing 10
Gene Aliases
Asb-10, BB137407
Chromosome Location
Chr.5: 24532685 - 24541047 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080444.4 NP_536692.1 5 - 6 1503 104
GenBank mRNA AF398971.1 - 5 - 6 1532 104
AK028538.1 - 5 - 6 1503 104
AK052929.1 - 5 - 6 1419 104
BC141311.1 - 5 - 6 1494 104
BC141313.1 - 5 - 6 1494 104

View Full Product Details