The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080452.3 | NP_536700.3 | 2 - 3 | 192 | 76 |
GenBank mRNA | AB055389.1 | - | 2 - 3 | 183 | 76 |
AK077219.1 | - | 3 - 4 | 361 | 76 | |
AK154131.1 | - | 2 - 3 | 186 | 76 | |
AK156412.1 | - | 2 - 3 | 165 | 76 | |
AK165956.1 | - | 2 - 3 | 183 | 76 | |
AK171699.1 | - | 2 - 3 | 183 | 76 | |
AK172432.1 | - | 2 - 3 | 165 | 76 | |
BC048377.1 | - | 2 - 3 | 176 | 76 | |
BC060503.1 | - | 2 - 3 | 168 | 76 |