Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aco2
Entrez Gene ID
11429
Gene Name
aconitase 2, mitochondrial
Gene Aliases
Aco-2, Aco3, D10Wsu183e
Chromosome Location
Chr.15: 81872463 - 81915137 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC004645.1 - 1 - 2 114 113

View Full Product Details