Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcolce
Entrez Gene ID
18542
Gene Name
procollagen C-endopeptidase enhancer protein
Gene Aliases
Astt-2, Astt2, P14, PCPE-1
Chromosome Location
Chr.5: 137605105 - 137611899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008788.2 NP_032814.2 8 - 9 1327 93
GenBank mRNA AB008548.1 - 8 - 9 1251 93
AK142615.1 - 8 - 9 1256 93
AK144392.1 - 8 - 9 1328 93
AK146829.1 - 8 - 9 1328 93
AK159940.1 - 8 - 9 1319 93
AK162524.1 - 7 - 8 2357 93
AK195180.1 - 1 - 2 85 93
X57337.1 - 8 - 9 1293 93

View Full Product Details