Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Etv4
Entrez Gene ID
18612
Gene Name
ets variant 4
Gene Aliases
AW414408, Pea-3, Pea3
Chromosome Location
Chr.11: 101769742 - 101785310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008815.2 NP_032841.2 7 - 8 800 128
GenBank mRNA AK144538.1 - 7 - 8 760 129
AK144590.1 - 7 - 8 753 128
AK144683.1 - 5 - 6 514 128
AK166136.1 - 7 - 8 736 128
DQ832277.1 - 6 - 7 505 128

View Full Product Details