The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009673.2 | NP_033803.1 | 4 - 5 | 527 | 118 |
GenBank mRNA | AK147740.1 | - | 4 - 5 | 306 | 118 |
AK152185.1 | - | 4 - 5 | 362 | 118 | |
AK153388.1 | - | 4 - 5 | 308 | 118 | |
AK153476.1 | - | 4 - 5 | 291 | 118 | |
BC003716.1 | - | 4 - 5 | 318 | 118 | |
CX218962.1 | - | 4 - 5 | 117 | ||
D63423.1 | - | 4 - 5 | 265 | 118 | |
U29396.1 | - | 4 - 5 | 284 | 118 |