The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081982.2 | NP_001075451.1 | 8 - 9 | 1572 | 101 |
NM_001297601.1 | NP_001284530.1 | 7 - 8 | 2121 | 101 | |
NM_010906.3 | NP_035036.1 | 7 - 8 | 1449 | 101 | |
GenBank mRNA | AK080940.1 | - | 1 - 2 | 121 | 101 |
AK158568.1 | - | 7 - 8 | 2124 | 101 | |
BC003766.1 | - | 8 - 9 | 1294 | 101 | |
U57636.1 | - | 6 - 7 | 1126 | 101 | |
Y07688.1 | - | 8 - 9 | 1574 | 101 | |
Y07689.1 | - | 7 - 8 | 1253 | 101 |