The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010918.2 | NP_035048.3 | 2 - 3 | 203 | 115 |
GenBank mRNA | AK035582.1 | - | 1 - 2 | 104 | 115 |
AK036649.1 | - | 2 - 3 | 168 | 115 | |
AK088056.1 | - | 2 - 3 | 142 | 115 | |
AK144214.1 | - | 2 - 3 | 114 | 115 | |
AK163916.1 | - | 2 - 3 | 216 | 115 | |
BC048414.1 | - | 1 - 2 | 137 | 115 | |
L04289.2 | - | 2 - 3 | 208 | 115 |