Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ntn3
Entrez Gene ID
18209
Gene Name
netrin 3
Gene Aliases
Ntn2l
Chromosome Location
Chr.17: 24203831 - 24209387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010947.3 NP_035077.1 2 - 3 1517 66
GenBank mRNA AF149094.1 - 2 - 3 1153 66
AF152418.1 - 2 - 3 1272 66
AK031199.1 - 2 - 3 1662 66
AK083904.1 - 2 - 3 1250 66
AK141856.1 - 2 - 3 1428 66
BC053038.1 - 2 - 3 1609 66
BC094362.1 - 2 - 3 1517 66

View Full Product Details