Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdcd4
Entrez Gene ID
18569
Gene Name
programmed cell death 4
Gene Aliases
D19Ucla1, Ma3, Tis
Chromosome Location
Chr.19: 53892231 - 53929861 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168491.1 NP_001161963.1 9 - 10 1292 95
NM_001168492.1 NP_001161964.1 9 - 10 1292 95
NM_011050.4 NP_035180.2 9 - 10 1380 95
GenBank mRNA AK134366.1 - 9 - 10 1381 95
AK172654.1 - 9 - 10 1248 95
BC055739.1 - 9 - 10 1227 95
CT010188.1 - 8 - 9 1090 95
D50465.1 - 8 - 9 1160 95
D86344.1 - 9 - 10 1176 95

View Full Product Details