Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pou2f3
Entrez Gene ID
18988
Gene Name
POU domain, class 2, transcription factor 3
Gene Aliases
Epoc-1, Oct-11a, Oct11, Otf-11, Otf11, Skin, Skin-1a, Skn-1a, Skn-li
Chromosome Location
Chr.9: 43123927 - 43205755 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011139.2 NP_035269.2 4 - 5 309 60
GenBank mRNA AK153697.1 - - 399 60
L14677.1 - 4 - 5 307 60
Z18537.1 - 4 - 5 310 60

View Full Product Details