Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bop1
Entrez Gene ID
12181
Gene Name
block of proliferation 1
Gene Aliases
AU020183, AW146150, D18861, Erb1p, Kiaa0124, mKIAA0124
Chromosome Location
Chr.15: 76452996 - 76477269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013481.1 NP_038509.1 8 - 9 1146 91
GenBank mRNA AK079627.1 - 8 - 9 1126 91
AK129058.1 - 8 - 9 1119 91
AK144095.1 - 8 - 9 1157 91
AK167110.1 - 8 - 9 1129 91
AK167225.1 - 8 - 9 1130 91
AK167829.1 - 8 - 9 1124 91
AK167878.1 - 8 - 9 1134 91
AK168127.1 - 8 - 9 1127 91
AK177135.1 - 1 - 2 69 91
AK180935.1 - 1 - 2 91
AK185180.1 - 1 - 2 69 91
AK188306.1 - 1 - 2 69 91
AK209200.1 - 2 - 3 268 91
AK217116.1 - 2 - 3 268 91
BC012693.1 - 8 - 9 1120 91
U77415.1 - 8 - 9 1146 91

View Full Product Details