The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013628.2 | NP_038656.1 | 13 - 14 | 1955 | 67 |
GenBank mRNA | BC108982.1 | - | 13 - 14 | 1933 | 67 |
BC108983.1 | - | 13 - 14 | 1933 | 67 | |
M58589.1 | - | 13 - 14 | 1955 | 67 | |
M69196.1 | - | 13 - 14 | 2061 | 67 | |
X57088.1 | - | 13 - 14 | 2067 | 67 |