The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048146.2 | NP_001041611.1 | 1 - 2 | 435 | 131 |
NM_001286507.1 | NP_001273436.1 | 1 - 2 | 435 | 131 | |
NM_013727.4 | NP_038755.1 | 1 - 2 | 435 | 131 | |
GenBank mRNA | AB007141.1 | - | 1 - 2 | 271 | 131 |
AI390224.1 | - | 1 - 2 | 435 | 131 | |
AK004992.1 | - | 1 - 2 | 414 | 131 | |
AK015942.1 | - | 1 - 2 | 256 | 131 | |
AK030076.1 | - | 1 - 2 | 267 | 131 | |
AK036102.1 | - | 1 - 2 | 342 | 131 | |
AK134714.1 | - | 1 - 2 | 338 | 131 | |
AK153477.1 | - | 1 - 2 | 399 | 131 | |
AK167783.1 | - | 1 - 2 | 379 | 131 | |
BC006851.1 | - | 1 - 2 | 330 | 131 |